Learn More
DWK Life Sciences Kimble™ Kontes™ TLC Cylindrical Developing Tanks
Cylindrical glass tanks for the development of 5 x 10cm (2 x 3.9 in. ) and 5 x 20cm (2 x 7.9 in. ) plates
Supplier:  DWK Life Sciences 4161701121
 View more versions of this product
Catalog No. K416170-1121
Only null left
Explore available promotions
Description
Description
Tanks provide maximum vapor phase saturation with a minimum amount of solvent
Specifications
Specifications
Glass
12/Pk.
Provides Maximum Vapor Phase Saturation with a Minimum Amount of Solvent
Cylindrical
Borosilicate Glass
Cylindrical TLC Developing Tank
5 L x 10 cm W Plate
Product Suggestions
Product Suggestions