Learn More
DWK Life Sciences Kimble™ Kontes™ Spargers for Tekmar™ Equipment
For use with Tekmar 2000 and 3000 Purge and Trap Concentrators and with 2016/2032 Autosamplers
Supplier:  DWK Life Sciences 5911002605
 View more versions of this product
Catalog No. K591100-2605
Only null left
Description
Description
Fritted Spargers
  • Recommended for potable water and wastewater samples
  • Fritless Spargers
    • Recommended for water, soils, polymers, pharmaceuticals and foods
    Specifications
    Specifications
    USP Type I, ASTM E438, Type I, Class A
    Potable Water and Wastewater
    Borosilicate Glass
    0.16 oz.
    5 mL
    Teledyne Tekmar™ 2000 and 3000 purge and trap concentrators and with the 2016 and 2032 autosamplers
    U
    Product Suggestions
    Product Suggestions