missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Round Bottom Three Neck Heavy Wall Distilling Flasks
With outer joints
Supplier: DWK Life Sciences 6060000624
Description
- Vertical side necks
Specifications
| 3.26 in. | |
| Borosilicate Glass | |
| Vertical | |
| 24/40 Center, 24/40 side | |
| 83 mm |
| 250 mL | |
| Standard taper outer joint | |
| 3 | |
| Round |