missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Pellet Pestle™ Microtubes
For use with Pellet Pestle
Supplier: DWK Life Sciences 7495100590
Description
For use with Pellet Pestle
- Choose autoclavable polypropylene or RNase-, DNase-, pyrogen-free material (manufactured under M5.5 [Class 10,000] cleanroom conditions)
- Individually wrapped
Chemical Identifiers
| Microtube | |
| Yes |
Specifications
| 0.01 oz. | |
| RNase-, DNase-, pyrogen-free | |
| Yes |
| 0.5 mL | |
| Microtube | |
| 0.01 oz., 0.5mL |