missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Incubation Flasks

Catalog No. p-122568
Change view
Click to view available options
Capacity (Metric):
10 mL
Closure Material:
Rubber
Material:
Borosilicate Glass
Includes:
w/o Sidearm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Material Material Includes
K8823000010 10 mL Rubber Borosilicate Glass w/o Sidearm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K8823000010 Supplier DWK Life Sciences Supplier No. 8823000010
Only null left

For metabolic studies of biological materials

Ideal for measuring C14O2 yields following incubation, in vitro incubations, insulin bioassays, persulfate oxidations, and distillation of any volatile acid or base. Also suitable for use with phosphate or bicarbonate buffers when acid or alkali is introduced at the end of a reaction.

  • Available with or without sidearm
  • Flask with sidearm enables syringe needle entry without disturbing established conditions

Order Info

Stoppers and center well sold separately.

Specifications

Closure Material Rubber
Capacity (Metric) 10 mL
Material Borosilicate Glass
For Use With (Application) For measuring C14O2 yields following incubation, in-vitro incubations, Insulin bioassays, Persulfate oxidations and distillation of any volatile Acid or base
Height (English) 2 in.
Height (Metric) 5 cm
Includes w/o Sidearm
Width (English) 1.2 in.
Width (Metric) 3.1 cm
Closure Type Stopper