missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Three-Valve Delivery Cap and Parts
Description
- Three on/off CTFE valves have 1/4-28 threads top and bottom
- Valve body is PTFE
- Valve stem is CTFE
- O-ring for valve stem and lower seal is TFE/propylene
Specifications
Specifications
| Color | Natural |
| Material | CTFE |
| No. per Case | 1 Cs. |
| For Use With (Equipment) | Ultra-ware™ standard three valve cap system |
| Product Type | Stem Valve |