Learn More
DWK Life Sciences Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Three-Valve Delivery Cap and Parts
Parts and accessories for standard three-valve cap system
Supplier:  DWK Life Sciences 9539030005
 View more versions of this product
Catalog No. K9539030005
Only null left
Description
Description
  • Three on/off CTFE valves have 1/4-28 threads top and bottom
  • Valve body is PTFE
  • Valve stem is CTFE
  • O-ring for valve stem and lower seal is TFE/propylene
Specifications
Chemical Identifiers
1 Cs.
Ultra-ware™ standard three valve cap system
Specifications
Blue
GL 45
Ultra-ware™ standard three valve cap system
1 Cs.
Screw Cap
Product Suggestions
Product Suggestions