Learn More
DWK Life Sciences Kimble™ Kontes™ HPLC Ultra-Ware™ Standard Three-Valve Delivery Cap and Parts
Parts and accessories for standard three-valve cap system
Supplier: DWK Life Sciences 9539030002
Description
- Three on/off CTFE valves have 1/4-28 threads top and bottom
- Valve body is PTFE
- Valve stem is CTFE
- O-ring for valve stem and lower seal is TFE/propylene
Chemical Identifiers
| 1 Cs. | |
| Ultra-ware™ standard three valve cap system |
Specifications
| Blue | |
| 1 Cs. | |
| Valve Handle | |
| Ultra-ware™ standard three valve cap system |