missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Heavy Wall Recovery Flasks with Standard Bead, Standard Taper Outer Joint

Catalog No. K2943000100
Change view
Click to view available options
Capacity (Metric):
100 mL
Standard Taper Joint:
14/20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Standard Taper Joint
K2943000100 100 mL 14/20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K2943000100 Supplier DWK Life Sciences Supplier No. 2943000100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KIMBLE™ KONTES™ Heavy Wall Recovery Flasks with Standard Bead, Standard Taper Outer Joint are designed for easy recovery of reaction products. Total contents are accessible with a spatula or policeman. Lower portion fits standard heating mantles.

  • Heavy wall single neck recovery flask with a standard taper outer joint
  • Designed for easy recovery of reaction products — total contents accessible with a spatula or policeman
  • Lower portion fits standard heating mantles
  • Even wall thickness and minimum runout make these flasks ideal for use with rotary evaporators
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Sold individually

Specifications

Capacity (Metric) 100 mL
Material Glass
For Use With (Application) Designed for easy recovery of reaction products
For Use With (Equipment) Rotary Evaporator
Neck Style Single
No. per Case 1/Cs.
Product Type Flask
Standard Taper Joint 14/20
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Round
Diameter (Metric) Outer 60 mm
Surface Treatment Non-treated
Show More Show Less