Learn More
DWK Life Sciences Kimble™ Kontes™ Flow Adapter Replacement Components for CHROMAFLEX™ Columns
Replacement HDPE Bed Supports for CHROMAFLEX™ Flow Adapters
Supplier:  DWK Life Sciences 4208112020
 View more versions of this product
Catalog No. K4208112020
Only null left
Description
Description
  • HDPE Bed Support
  • 20μm HDPE Bed Support
  • 1.0cm, 2.5cm and 4.8cm options
  • Case of 10
Specifications
Chemical Identifiers
HDPE
Specifications
2.5 cm
HDPE
Product Suggestions
Product Suggestions