missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Filtration Assemblies: Borosilicate Glass Funnels for 47mm Assemblies

Accessory for 47mm Assemblies

Supplier:  DWK Life Sciences 9537610000

 View more versions of this product

Catalog No. K9537610000


Only null left

Description

Description

  • Accessory/ Replacement for: K953755-0000, K953805-0000, K953855-1047, K953825-0000, K953835-0000, K953845-0000

Specifications

Chemical Identifiers

Borosilicate Glass
47 mm Assemblies

Specifications

100 mL
Borosilicate Glass
47 mm Assemblies
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions