Learn More
DWK Life Sciences Kimble™ Kontes™ Erlenmeyer Flasks
Used to measure, mix, and store liquids
Supplier: DWK Life Sciences 6170000724
Description
Used to measure, mix, and store liquids
- Single neck flask
- Has standard taperouter joint
Specifications
| 1000 mL | |
| 9.05 in. | |
| 24/40 |
| Borosilicate Glass | |
| 23 cm | |
| Narrow |