missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Distilling Two-Neck Round Bottom Heavy-Wall Flasks
Side neck is angled or vertical
Supplier: DWK Life Sciences 6050201824
Description
- Heavy wall reduces breakage.
Specifications
| 6.33 in. | |
| Borosilicate Glass | |
| 2 | |
| Round |
| 2000 mL | |
| Angled | |
| 24/40 Center, 24/40 side | |
| 160 mm |