missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Distilling Two-Neck Round Bottom Heavy-Wall Flasks
Side neck is angled or vertical
Supplier: DWK Life Sciences 6050200210
Description
- Heavy wall reduces breakage.
Specifications
| 2.36 in. | |
| Borosilicate Glass | |
| Angled | |
| 24/40 Center, 10/30 side | |
| 60 mm |
| 100 mL | |
| Standard taper outer joint | |
| 2 | |
| Round |