Learn More
DWK Life Sciences Kimble™ Kontes™ Allihn Condensers with Straight Outlet Tube
For refluxing applications including Soxhlet extractions
Supplier:  DWK Life Sciences 4560000021
 View more versions of this product
Catalog No. K456000-0021
Only null left
Description
Description
  • Straight outlet tube at top Water-cooled standard taper inner drip joint at bottom
  • Accept No. 2 hose connections, except for 350mm size, which accepts No. 3
Specifications
Specifications
Straight outlet tube, water cooled standard taper inner drip joint at bottom
34/45
27.5 cm
10.8 in.
Borosilicate Glass
0.37 in.
7.4 in.
Refluxing operations, including soxhlet extractions
Allihn Condenser
19 cm
0.952 cm
USP Type I and ASTM E438 Type I, Class A
Product Suggestions
Product Suggestions