missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ 1000 mL Four Vertical Neck Round Bottom Flask
Description
- Manufactured from 33 expansion, low extractable borosilicate glass
- Conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
| Capacity (Metric) | 1000 mL |
| Side Arm Joint | 24/40 mm |
| Socket Joint | 24/40 mm |
| Diameter (Metric) Outer | 130 mm |