missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences KIMBLE™ KIMAX™ Iodine Determination Flasks with PTFE Stopper
Description
For analysis of dry shellac, varnishes, oils, fatty acids
- Used for the analysis of dry shellac and shellac varnishes with Wijs iodine monochloride solution, and to determine the total iodine value of oils, fatty acids, and bodied oils with Rosenmund-Kuhnhenn reagent
- PTFE standard taper stopper is included
- Durable matte-finish ceramic enamel marking spot takes writing and erases easily
- Designed to ASTM Specification E1404, Type II, Class 3 requirements
Compliance
Designed from ASTM E 1404, Type II, Class 3 requirements
Specifications
Specifications
| Closure Material | PTFE |
| Diameter (English) Outer | 4.01 in. |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass |
| Height (English) | 7.67 in. |
| Height (Metric) | 19.5 cm |
| Product Type | Erlenmeyer Flask |
| Writing Surface | Matte Finish Ceramic Enamel Marking |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Show More |