Learn More
Kimble Kimax Graduated Culture Tubes
Supplier:  DWK Life Sciences 4507110
Catalog No. K4507110
Only null left
Description
Description
Specifications
Specifications
USP Type I, ASTM E 438 Type I, Class A, APHA, E 982, Type II
150 mm
Single-use
White Ceramic Enamel Marking Spot
16 mm
10 mL
Borosilicate Glass
For testing of milk for the presence of bacteria by the methylene blue reductase test
Yes
Product Suggestions
Product Suggestions