missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ KIMAX™ Burets with Threaded Closures Replacement PTFE Stopcock Assembly

Catalog No. 10310166
Change view
Click to view available options
Plug Bore:
2.5 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
10310166 2.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10310166 Supplier DWK Life Sciences Supplier No. 41007F2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The KIMBLE™ KIMAX™ Burets with Threaded Closures Replacement PTFE Stopcock Assembly features a PTFE LUBRI-FLO™ stopcock with a 2.8 mm bore, designed for use with 29053H and 17121H burets. Nylon threaded closure (15-415 GPI) ensures secure attachment.

  • PTFE LUBRI-FLO™ stopcock, all surfaces in contact with liquid are made of PTFE
  • 2.8 mm bore plug for precise flow control
  • Compatible with Kimble™ KIMAX™ 17121H and 29053H threaded-closure burets
  • Nylon threaded closure (15-415 GPI) provides secure attachment to compatible burets
  • Tip should be wetted before installation for easier assembly
  • Sold individually (Case Qty: 1)

Specifications

Type Replacement Stopcock
Plug Bore 2.5 mm
Material PTFE
Closure Size 15-415
For Use With (Application) Burets
For Use With (Equipment) 29053H and 17121H Burets
Plug Material PTFE