missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ White Polypropylene Closure with Pulp-Backed Aluminum Foil

Catalog No. 03340130
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03340130 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03340130 Supplier DWK Life Sciences Supplier No. 7452115425
Only null left

Top suitable for marking

  • For scintillation vial
  • White polypropylene
  • Pulp-backed aluminum foil cap liner
  • For use with strong oxidizing agent
  • GPI thread finish: 15-425

Specifications

Material (Liner) Aluminum Foil/Pulp
Color White
Material Polypropylene
For Use With (Application) Various strong oxidizing agent mixtures