Learn More
DWK Life Sciences Kimble™ Closures for 20mL Glass and Plastic Scintillation Vials
Catalog No. 033378
Change view
Click to view available options
Material:
Urea
Material (Liner):
Polyethylene
PTFE/Rubber
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Material (Liner)
033378 Urea Polyethylene
03340131 Urea PTFE/Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 033378 Supplier DWK Life Sciences Supplier No. 7452522400
Only null left
Durable screw thread closures
  • For 20mL Fisherbrand™ and Kimble scintillation vials, except 03-340-4B and 03-340-129. GPI 22-400 thread.
Specifications
Material (Liner) Polyethylene
Color White
Material Urea