Learn More
DWK Life Sciences Kimble™ Rim Top Mark-M™ Borosilicate Glass Culture Tubes with Marking Labels
Permanent ceramic label, fused to the glass located 3/4 in. from the open end
Supplier:  DWK Life Sciences 60B12BZW
 View more versions of this product
Catalog No. 22378844
Only null left
Description
Description
Permanent ceramic label, fused to the glass located 3/4 in. from the open end
  • Borosilicate glass
  • Available in a variety of colors
  • Sturdy, uniform bottoms and consistent lengths
Specifications
Chemical Identifiers
USP Type I, ASTM E 438 Type I, Class B
Single-use
Marking Spot
Borosilicate Glass
1.37 in. permanent ceramic vertical marking label
Yes
Specifications
USP Type I, ASTM E 438 Type I, Class B
5 mL
Borosilicate Glass
1.37 in. permanent ceramic vertical marking label
Yes
White
75 mm
Single-use
Marking Spot
12 mm
Product Suggestions
Product Suggestions