Learn More
DWK Life Sciences Kimble™ Kontes™ Tube For Potter-Elvehjem Tissue Grinder
Catalog No. K8855120021
Change view
Click to view available options
Size:
20
21
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
K8855120021 21
K8855120020 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K8855120021 Supplier DWK Life Sciences Supplier No. 8855120021
A graduated tube with markings fused onto the glass. Range of tube sizes: Size 20 (4mL), Size 21 (10mL) and Size 22 (55mL).
  • Graduations are fused onto the glass
  • Clearance between pestle and tube
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Size 21
Product Type Tissue Grinder Tube
For Use With (Equipment) Potter-Elvehjem tissue grinder