Learn More
DWK Life Sciences Kimble™ Duall™ All-Glass Tissue Grinder Accessory, Grinding Tube
Catalog No. K8854620025
Change view
Click to view available options
Includes:
With plastic safety coating
Without plastic safety coating
Size:
20
21
22
23
24
25
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
K8854620025 With plastic safety coating 25
K8854520021 Without plastic safety coating 21
K8854520022 Without plastic safety coating 22
K8854520020 Without plastic safety coating 20
K8854620021 With plastic safety coating 21
K8854620020 With plastic safety coating 20
K8854620022 With plastic safety coating 22
K8854620023 With plastic safety coating 23
K8854620024 With plastic safety coating 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. K8854620025 Supplier DWK Life Sciences Supplier No. 8854620025
Improves grinding efficiency
  • Tubes sizes from 21 to 25
  • Available with or without coating
Specifications
Size 25
Material Plastic
Product Type Tissue Grinder Tube
For Use With (Application) Improves grinding efficiency
Includes With plastic safety coating
For Use With (Equipment) Duall™ all Glass tissue grinders