missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Titeseal™ 4mL Shell Vial Needle Closures

Catalog No. 0334018E
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
0334018E White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0334018E Supplier DWK Life Sciences Supplier No. 738353

For use with Kimble-Chase Kontes Titeseal 4mL Shell Vials

  • Polyethylene plug-style needle closures

Specifications

Color White
Material Polyethylene
Closure Type Plug Style Needle
Quantity 500 Pk.