Learn More
DWK Life Sciences Kimble™ Kontes™ Three Neck Reaction Flask
Catalog No. K6125000021
Change view
Click to view available options
Standard Taper Joint:
24/40
Catalog No. Standard Taper Joint
K6125000021 24/40
1 options
Catalog No. K6125000021 Supplier DWK Life Sciences Supplier No. 6125000021
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with cylindrical and spherical flasks having the same I.D.
  • Flat ground flange
Specifications
Diameter (Metric) Inner Flange 10.16 cm
Material Borosilicate Glass
For Use With (Equipment) Cylindrical and spherical flasks
No. of Positions 3
Product Type Flask, Reaction, Kontes™ three neck
Standard Taper Joint 24/40