missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Micro-Scale Tissue Grinder, All Glass

Catalog No. K8854700000
Change view
Click to view available options
Capacity (Metric):
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
K8854700000 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K8854700000 Supplier DWK Life Sciences Supplier No. 8854700000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

With ground glass pestle

 

  • Manufactured from borosilicate glass 3.3
  • Designed for dispersing small quantities of tissue in buffer solution
  • Working capacity is 0.2 mL 
  • All-glass tube and pestle
  • PTFE bearing prevents misalignment and avoids aerosol formation

 

Specifications

Diameter (English) Shaft 0.11 in.
Diameter (Metric) Shaft 3mm
Capacity (English) 0.006 oz.
Capacity (Metric) 0.2 mL
Material Ground Glass
Product Type Tissue Grinder
For Use With (Application) Designed for dispersing small quantities of tissue in Buffer solution
Length (English) Pestle 2.9 in.
Length (Metric) Pestle 75mm
Length (English) Tube 1.2 in.
Length (Metric) Tube 33mm
Tube Diameter (English) Outside 0.31 in.
Tube Diameter (Metric) Outside 8mm
Quantity 6/Cs.
Show More Show Less