missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Packing Reservoir for CHROMAFLEX™ Columns
Description
- Fits Standard Column
- Each packing reservoir is designed for easy mounting on the CHROMAFLEX™ columns
- The capacities are matched to take the amount of gel slurry required to pack the longest column
- The packing reservoirs may also be used as buffer reservoirs for gravity flow chromatography
- 1.0cmID column has nominal volume of 150mL
- 2.5cmID column has nominal volume of 500mL
- Temperature Range 0 to 50°C
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Packaging | End fitting has 1/4-28 threads for easy connection to pumps |
| Quantity | 150 mL |
| Type | Packing Reservoir |
| Diameter (Metric) | 1 cm |