Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Nut for 1/8 in. Tubing
Catalog No. K4208210018
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
K4208210018 PTFE
1 options
Catalog No. K4208210018 Supplier DWK Life Sciences Supplier No. 4208210018
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
  • ETFE Flangeless Nut
  • 1/4-28 Thread, for 1/8 in. and 1.8mm OD Tubing
  • Use with the 420822-Series Flangeless Ferrules
  • Case of 10
  • Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
Specifications
For Use With (Equipment) HPLC cap system
Type Nut
Material PTFE
Quantity 1 Cs.