Learn More
DWK Life Sciences Kimble™ Kontes™ Glass Dispersion Tubes
Catalog No. K9565000025
Change view
Click to view available options
Tube, EC Porosity
Catalog No.
K9565000025 Tube, EC Porosity
1 options
Catalog No. K9565000025 Supplier DWK Life Sciences Supplier No. 9565000025
With hollow fritted cylinder at the end of tube
  • Extra coarse porosity cylinder
Specifications
Diameter (Metric) Inner 8 mm
Height (Metric) 250 mm
Product Type Tube