Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Flow Control Adapter with 24/40 ST Joint
The flow control adapter features Kimble™ Kontes™ Hi-Vac™ PTFE plug designed with a precision external thread for fine control.
Supplier:  DWK Life Sciences 4205752440
Catalog No. K4205752440
Only null left
Explore available promotions
Description
Description
  • The flow control adapter features Kontes Hi-Vac™ PTFE plug designed with a precision external thread for fine control
  • The accurately controlled valve seat and the extended tip on the PTFE plug allow semi-needle valve control for liquids
  • The FKM o-ring is shielded from direct exposure to liquids within the system by PTFE ring seals
Specifications
Specifications
Flow Control Adapter
1 Ea.
Joint
Product Suggestions
Product Suggestions