missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Flow Adapter for 2.5 CHROMAFLEX™ Standard Columns
Description
- Fits Standard Column
- Flow adapters are recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations
- Each flow adapter is supplied with a 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, blue lock ring, and 5 feet (1.5 meters) of FEP tubing
- 2.5cm column ID
- Temperature Range 0 to 50°C
- Tubing OD 1/16 in.
- Use of aggressive organic solvents requires the PTFE bed supports listed as accessories and FFKM o-rings sizes 010, 116 or 222 (not available from Kimble)
- Case of 1
Specifications
Specifications
| Type | Flow Adapter |
| Diameter (English) Outer | 1/16 in. |
| Includes | 20μ porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 feet (1.5 meters) of FEP PTFE tubing |
| For Use With | Fits Column ID : 2.5cm |
| Material | PTFE |