Learn More
DWK Life Sciences Kimble™ Kontes™ Flow Adapter for 4.8 CHROMAFLEX™ Standard Columns
Catalog No. K4208360040
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
K4208360040 PTFE
1 options
Catalog No. K4208360040 Supplier DWK Life Sciences Supplier No. 4208360040
Flow adapters improve chromatogram resolution by delivering the sample and buffer directly to the top of the gel bed.  This protects the gel bed from disruption during sample loading and eliminates any dead volume above the gel bed.
  • Fits Standard Column
  • Flow adapters are recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations
  • Each flow adapter is supplied with a 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, blue lock ring, and 5 feet (1.5 meters) of FEP tubing
  • 4.8cm column ID
  • Temperature Range 0 to 50°C
  • Tubing OD 1/8 in.
  • Use of aggressive organic solvents requires the PTFE bed supports listed as accessories and FFKM o-rings sizes 010, 116 or 222 (not available from Kimble)
  • Case of 1
Specifications
Type Flow Adapter
Diameter (English) Outer 1/8 in.
Includes 20μ porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 feet (1.5 meters) of FEP PTFE tubing
For Use With Fits Column ID : 4.8cm
Material PTFE