Learn More
DWK Life Sciences Kimble™ Kontes™ Flow Adapter for 1.0 CHROMAFLEX™ Standard Columns
Catalog No. K4208360000
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
K4208360000 Stainless Steel
1 options
Catalog No. K4208360000 Supplier DWK Life Sciences Supplier No. 4208360000
Includes
20μm porosity HDPE bed support, TFE/propylene O-ring seal, stainless-steel adjusting rod, PTFE body and 5 ft. (1.5m) of PTFE tubing.
Flow adapters improve chromatogram resolution by delivering the sample and buffer directly to the top of the gel bed.  This protects the gel bed from disruption during sample loading and eliminates any dead volume above the gel bed.
  • Fits Standard Column
  • Flow adapter is recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations
  • Each flow adapter is supplied with a 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 feet (1.5 meters) of FEP tubing
  • 1.0cm column ID
  • Temperature Range 0 to 50°C
  • Tubing OD 1/16 in.
  • Use of aggressive organic solvents requires the PTFE bed supports listed as accessories and FFKM o-rings sizes 010, 116 or 222 (not available from Kimble)
  • Case of 1
Specifications
Diameter (Metric) Inner 1 cm
Material Stainless Steel