Learn More
DWK Life Sciences KIMBLE™ KONTES™ FlexColumn™ Three-way Luer Stopcock
Description
Heavy-duty nylon
- Three-way stopcock valve with heavy-duty nylon body and HDPE valve plug
- Three-way flow control with locking fittings on all ports
- 2 female Luer inlets to 1 male Luer lock outlet configuration
- Supplied in pack of 10
Specifications
Specifications
| Type | 3-way Stopcock |
| Material | Nylon |
| For Use With (Equipment) | FlexColumn™ |
| Includes | Locking fittings on all ports |
| Plug Material | HDPE |
| Outlet Port | Female Luer to Male Luer |
| Connectors | Female Luer to Male Luer |