Learn More
DWK Life Sciences Kimble™ Kontes™ Flex-Column Economy Luer Adapter and Tubing
Catalog No. KH906690005
Change view
Click to view available options
For Use With:
CHROMAFLEX™ columns
Catalog No. For Use With
KH906690005 CHROMAFLEX™ columns
1 options
Catalog No. KH906690005 Supplier DWK Life Sciences Supplier No. H906690005
Chemically inert. Suitable for low-pressure applications.
  • Adapter consists of 5 ft. (1.5m) length of 0.038 in. I.D. (approx. 1mm) PTFE tubing with integral CTFE locking fitting.
Specifications
Diameter (English) Outer 1/16 in.
Includes Tubing, integral F-Luer Fitting
For Use With CHROMAFLEX™ columns
Overall Length (English) 5 ft.
Material PTFE (Tubing), CTFE (Luer)