missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Flex-Column Economy Columns Accessory: Polypropylene Reservoirs

Catalog No. K4204050700
Change view
Click to view available options
Volume (Metric):
100 mL
700 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
K4204050700 700 mL
K4204050100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K4204050700 Supplier DWK Life Sciences Supplier No. 4204050700
Only null left

For use with Kimble-Chase Kontes Flex-Column Economy Columns

  • Available in two sizes: 100mL and 700mL
  • 100mL: fits 0.7, 1.0, and 1.5cm diameter Flex-Column columns
  • 700mL: fits 2.5cm columns
  • For Flex-Column Economy Columns, see cat. no. K420400-1015

Specifications

Type Packing Reservoir
Volume (Metric) 700 mL
For Use With 2.5cm I.D. FlexColumn™ economy column for initial column packing and as a small Buffer reservoir
Material Polypropylene