missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Flex-Column Economy Columns Accessory: Polypropylene Reservoirs
Description
For use with Kimble-Chase Kontes Flex-Column Economy Columns
- Available in two sizes: 100mL and 700mL
- 100mL: fits 0.7, 1.0, and 1.5cm diameter Flex-Column columns
- 700mL: fits 2.5cm columns
- For Flex-Column Economy Columns, see cat. no. K420400-1015
Specifications
Specifications
| Type | Packing Reservoir |
| Volume (Metric) | 100 mL |
| For Use With | 0.7, 1 and 1.5cm I.D. FlexColumn™ economy column for initial column packing and as a small Buffer reservoir |
| Material | Polypropylene |