Learn More
DWK Life Sciences KIMBLE™ KONTES™ Filtration Assemblies: Flasks With Side Arm, No. 2 Hose Connection
Catalog No. K9537604000
Change view
Click to view available options
Capacity (Metric):
125 mL
1000 mL
2000 mL
4000 mL
Standard Taper Joint:
5
8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Standard Taper Joint
K9537604000 4000 mL 8
K9537100000 125 mL 5
K9537600000 1000 mL 8
K9537602000 2000 mL 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. K9537604000 Supplier DWK Life Sciences Supplier No. 9537604000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KIMBLE™ KONTES™ Filtration Assemblies: Flasks With Side Arm, No. 2 Hose Connection are borosilicate glass filtration flasks with a #2 hose connection for 0.25 in. (6mm) ID tubing, conforming to USP Type I and ASTM E438, Type I, Class A requirements.
DWK Life Sciences Kimble Kontes Filtration Assemblies: Flasks With Side Arm, No. 2 Hose connection, Connector Types: No. 2 Hose Connection, Borosilicate Glass, Description: Filtration Flask
  • Borosilicate glass filtration flask with side arm hose connection
  • #2 hose connection designed for 0.25 in. (6mm) ID flexible tubing
  • 125mL flask features a No. 5 stopper joint; 1000mL, 2000mL, and 4000mL flasks feature a No. 8 stopper joint
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Suitable for analysis of particulate or microbiological contamination
  • A second flask should be connected between the filtering flask and the vacuum source to prevent accidental entry of filtrate into the vacuum line or pump
Order Info
Does not include rubber stopper and hose
Specifications
Capacity (Metric) 4000 mL
Material Borosilicate Glass
Includes No. 2 Hose Connection
Product Type Filtration Flask
Standard Taper Joint 8
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Flat
Surface Treatment Non-treated
Quantity 1/Cs.
Show More Show Less