Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE End Fitting for Chromaflex Columns
Replacement PTFE End Fitting with Bed Support for CHROMAFLEX™ and FLEX-COLUMN™ columns
$134.68 - $164.36
Specifications
Type End Fitting
Includes Bed supports
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
K420832-2100
View Documents
DWK Life Sciences
4208322100
10 mm N/A
Only null left
K420832-2110
View Documents
DWK Life Sciences
4208322110
250 mm N/A
Only null left
K420832-2120
View Documents
DWK Life Sciences
4208322120
480 mm N/A
Only null left
Description
Description
Replacement PTFE End Fitting with Bed Support for CHROMAFLEX™ and FLEX-COLUMN™ columns
  • PTFE Ending Fitting with Bed Support
  • 1.0cm, 2.5cm and 4.8cm options
  • Requires attachment with CTFE adapter 420804-0001, not included
Specifications
Specifications
End Fitting
Bed supports
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'