Learn More
DWK Life Sciences Kimble™ Kontes™ 150mm Condensor for Solvent Recovery System
This Condensor is a replacement component of a three-position linear solvent recovery system intended for use with Kuderna Danish evaporative concentrators.
Supplier:  DWK Life Sciences 5455041518
Numéro de catalogue. K5455041518
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • System Quantity 3
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Spécifications
Spécifications
Condenser
3-position Linear Solvent Recovery System
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS