Learn More
DWK Life Sciences Kimble™ Kontes™ Column with Standard Taper Joint
Gravity column manufactured from medium wall tubing with PTFE stopcock.
Supplier: DWK Life Sciences 4205500245
Description
- Glass frit (40-60 micron porosity) at the lower end to act as a support for column packing material
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
- Outer joint ( 24/40) at top permits connection of addition funnel or other reservoir
Specifications
| 22 mm | |
| 420570 Series reservoir and 420575 series flow control adapters | |
| 250 mL | |
| Glass |
| Gravity Chromatography Column | |
| 40 to 60 μm | |
| 550 mm | |
| Fritted disc, 2mm bore PTFE stopcock plug and 24/40 standard taper upper joint |