Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Cylindrical TLC Developing Tanks
Cylindrical glass tanks for the development of 1 x 3 in. to 10 x 20cm plates.
Supplier:  DWK Life Sciences 4161701020
 View more versions of this product
Catalog No. K4161701020
Only null left
Description
Description
  • These tanks provide maximum vapor phase saturation with a minimum amount of solvent
  • Polyethylene or glass cap included
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Chemical Identifiers
Cylindrical TLC Developing Tank
Provides Maximum Vapor Phase Saturation with a Minimum Amount of Solvent
Cylindrical
Specifications
Glass
Cylindrical TLC Developing Tank
10 x 20 cm Plate Size
Cylindrical
Glass
Provides Maximum Vapor Phase Saturation with a Minimum Amount of Solvent
10 H x 5 cm I.D. Plate
Product Suggestions
Product Suggestions