Learn More
DWK Life Sciences Kimble™ KIMAX™ Squibb Pear-Shaped Sep. Funnel with PTFE-Plug Stopcock, standard taper PE Stopper
Closed-bottom, linear high-density polyethylene stopper in standard taper ground neck
$624.00 - $1,824.14
Specifications
Material Borosilicate Glass
Certifications/Compliance ASTM E1096, Type IV
Length (Metric) Stem 60mm
Stopper Material HDPE
Product Type Squibb Separatory Funnel
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (Metric) Stopcock Stopper Number Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Stopcock Stopper Number Quantity Price Quantity & Availability  
1043711F
View Documents Promotion Details
DWK Life Sciences
29049F1000
1000 mL 4; PTFE 27 Each
Each for $867.32
Case of 2 Each for $1,734.64
Only null left
1043711E
View Documents Promotion Details
DWK Life Sciences
29049F500
500 mL 4; PTFE 27 Each
Each for $624.00
Case of 4 Each for $2,496.00
Only null left
1043711G
View Documents Promotion Details
DWK Life Sciences
29049F2000
2000 mL 4; PTFE 38 Each
Each for $1,252.79
Case of 2 Each for $2,505.58
Only null left
1043711D
View Documents Promotion Details
DWK Life Sciences
29049F250
250 mL 4; PTFE 22 2 Pack
Pack of 2 Each for $1,090.25
Case of 2 Pack for $2,180.50
Only null left
1043711C
View Documents Promotion Details
DWK Life Sciences
29049F125
125 mL 2; PTFE 22 2 Pack
Case of 4 Each for $1,824.14
Only null left
Description
Description
Closed-bottom, linear high-density polyethylene stopper in standard taper ground neck
Lower stem made of large-diameter tubing for liquid to drain out completely when stopcock is closed.
Order Info
Stem lengths, 60mm.
Compliance
Designed from ASTM™ E 1096-86.
Specifications
Specifications
Borosilicate Glass
60mm
Squibb Separatory Funnel
ASTM E1096, Type IV
HDPE