Learn More
DWK Life Sciences Kimble™ KIMAX™ Serialized/Certified Cylindrical Microflasks
Catalog No. 208145F
Change view
Click to view available options
Capacity (Metric):
1 mL
2 mL
5 mL
10 mL
25 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
208145F 10 mL
10212H 5 mL
10212L 25 mL
10212K 2 mL
10212J 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 208145F Supplier DWK Life Sciences Supplier No. 28017A10
Calibrated To Contain
Calibrated To Contain
  • Permanently marked with individual serial number
  • Individually calibrated and certified against equipment whose calibration is traceable to NIST™
  • Cylindrical body for better mixing and draining
  • Easier to withdraw samples using pipets
  • Wide base provides stability
  • Marking spot
  • Ground glass stopper
  • 1 and 2mL sizes come with standard taper 8 stopper; 10mL size with standard taper 9
stopper
Compliance
Designed from requirements of ASTM™ E 237, Class A serialized.
Specifications
Capacity (Metric) 10 mL