Learn More
DWK Life Sciences Kimble™ High Temperature Cap
Catalog No. 02542350
Change view
Click to view available options
Thread Size (Metric):
GL 45
Catalog No. Thread Size (Metric)
02542350 GL 45
1 options
Catalog No. 02542350 Supplier DWK Life Sciences Supplier No. DWK14395H452
High-quality, chemical-resistant cap and liner withstands up to 180°C
  • Provides good chemical resistance to alcohols, ethers, hydrocarbons, and dilute or strong acids
  • Fits all GL 45 bottles and flasks
  • Comes in red and includes liner
  • Made from Polybuthyleneteraphthalate (PBT) cap with Polytetrafluoroethylene (PTFE) faced silicone liner
Specifications
Color Red
Material PBT
No. per Case 10 Cs.
Thread Size (Metric) GL 45
Liner Material PTFE/Silicone
Neck Finish 43 mm
Product Type High Temperature Cap