missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
$439.30 - $594.37
Specifications
| Capacity (English) | 0.05 oz. |
|---|---|
| For Use With (Application) | For animal tissues and organs, plant materials, insects, DNA, RNA, Proteins, yeasts and Enzymes |
| Autoclavable | Autoclavable |
| Color | Clear |
| Includes | PP tube, POM pestle |
| Catalog Number | Mfr. No. | Packaging | Sterility | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging | Sterility | Price | Quantity | ||||
|
K7496250010
|
DWK Life Sciences
7496250010 |
Bulk | Non-sterile |
Case of 50 Each for $439.30
|
|
||||
|
K7496250030
|
DWK Life Sciences
7496250030 |
Single pack | Sterile |
Case of 50 Each for $594.37
|
|
||||
Description
Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials
- Clear polypropylene tube
- Polyacetal pestle with molded-in shaft guard
- Abrasive surface (pestle tip and inner tube area)
- Available in Sterile and non-sterile versions
- Capacity: 1.5mL
For animal tissues and organs, plant materials, insects, DNA, RNA, proteins, yeasts and enzymes
Specifications
| 0.05 oz. | |
| Autoclavable | |
| PP tube, POM pestle | |
| 0.05 oz., 1.5mL |
| For animal tissues and organs, plant materials, insects, DNA, RNA, Proteins, yeasts and Enzymes | |
| Clear | |
| 1.5 mL |