missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer

Catalog No. K7496250010
Change view
Click to view available options
Packaging:
Bulk
Single pack
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Sterility
K7496250010 Bulk Non-sterile
K7496250030 Single pack Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K7496250010 Supplier DWK Life Sciences Supplier No. 7496250010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials

  • Clear polypropylene tube
  • Polyacetal pestle with molded-in shaft guard
  • Abrasive surface (pestle tip and inner tube area)
  • Available in Sterile and non-sterile versions
  • Capacity: 1.5mL

For animal tissues and organs, plant materials, insects, DNA, RNA, proteins, yeasts and enzymes

Specifications

Capacity (English) 0.05 oz.
For Use With (Application) For animal tissues and organs, plant materials, insects, DNA, RNA, Proteins, yeasts and Enzymes
Autoclavable Autoclavable
Color Clear
Includes PP tube, POM pestle
Capacity (Metric) 1.5 mL
Packaging Bulk
Sterility Non-sterile