Learn More
DWK Life Sciences Kimble™ BEAKERplus™ Combination Beakers and Flasks
Combination design to reduce splashing during agitation with funnels or filters
Supplier:  DWK Life Sciences 14035500
 View more versions of this product
Catalog No. 02555104
Only null left
Description
Description
  • Made from KG-33 borosilicate glass
  • Graduated
  • Wide fluted rim
  • Pour spout
  • Narrow recessed neck
  • Chemical and heat resistant
Specifications
Chemical Identifiers
Borosilicate Glass
Yes
Specifications
2.75 in.
500 mL
7.007 in.
Yes
70 mm
Borosilicate Glass
17.8 cm
Product Suggestions
Product Suggestions