missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials with Attached White Rubber Lined Closures
Attached closed top black phenolic closures
$351.37 - $711.67
Specifications
| Material (Liner) | Rubber |
|---|---|
| Color | Amber |
| Material | Borosilicate Glass |
| Thread Style | Screw |
| Closure Material | Phenolic |
| Catalog Number | Mfr. No. | Closure Type | Volume (English) | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Closure Type | Volume (English) | Price | Quantity | ||||
|
14-957-256
|
N/A | Screw Cap | 0.37 dr. |
N/A
|
|
||||
|
14-957-255
|
N/A | Screw Cap (Closed-top) | 1 dr. |
N/A
|
|
||||
|
14-957-258
|
N/A | Screw Cap (Closed-top) | 5.4 dr. |
N/A
|
|
||||
|
14-957-257
|
N/A | Screw Cap (Closed-top) | 2.16 dr. |
N/A
|
|
||||
Description
Attached closed top black phenolic closures
- Manufactured from amber 51 expansion borosilicate glass
- Closures with white rubber liners
- Vials packaged in corrugated trays with partitions
Specifications
| Rubber | |
| Borosilicate Glass | |
| Phenolic |
| Amber | |
| Screw | |
| USP Type I |